1-Brom-4-pentylbenzol

Lieferant: Thermo Scientific

Synonyme: 4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol

BTB09867.10 BTB09867.50
MAYBBTB09867.10EA 0 EUR
MAYBBTB09867.10 MAYBBTB09867.50
1-Brom-4-pentylbenzol
1-Brom-4-pentylbenzol
Formel: C₁₁H₁₅Br
Molecular Weight: 227.14 g/mol
MDL: MFCD00061113
CAS-Nummer: 51554-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR