4-Ethylbenzoesäure

Lieferant: Thermo Scientific

BTB09873.1 BTB09873.10 BTB09873.50
MAYBBTB09873.1EA 0 EUR
MAYBBTB09873.1 MAYBBTB09873.10 MAYBBTB09873.50
4-Ethylbenzoesäure
4-Ethylbenzoesäure
Formel: C₉H₁₀O₂
Molecular Weight: 150.18 g/mol
MDL: MFCD00002570
CAS-Nummer: 619-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR