4-Ethylbenzoesäure
Lieferant: Thermo Scientific
BTB09873.1
BTB09873.10
BTB09873.50
MAYBBTB09873.1EA
0
EUR
MAYBBTB09873.1
MAYBBTB09873.10
MAYBBTB09873.50
4-Ethylbenzoesäure
4-Ethylbenzoesäure
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 g/mol |
MDL:
MFCD00002570 CAS-Nummer: 619-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...