4-Butylbenzoylchlorid
Lieferant: Thermo Scientific
Synonyme:
4-Butylbenzol-1-carbonylchlorid
BTB09878.1
BTB09878.10
BTB09878.50
MAYBBTB09878.1EA
0
EUR
MAYBBTB09878.1
MAYBBTB09878.10
MAYBBTB09878.50
4-Butylbenzoylchlorid
4-Butylbenzoylchlorid
Formel:
C₁₁H₁₃ClO Molecular Weight: 196.68 g/mol Siedepunkt: 155 Dichte: 1.05 g/cm³ (20 °C) |
MDL:
MFCD00000699 CAS-Nummer: 28788-62-7 EINECS: 249-224-2 UN: 3265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...