4-Butylbenzoylchlorid

Lieferant: Thermo Scientific

Synonyme: 4-Butylbenzol-1-carbonylchlorid

BTB09878.1 BTB09878.10 BTB09878.50
MAYBBTB09878.1EA 0 EUR
MAYBBTB09878.1 MAYBBTB09878.10 MAYBBTB09878.50
4-Butylbenzoylchlorid
4-Butylbenzoylchlorid
Formel: C₁₁H₁₃ClO
Molecular Weight: 196.68 g/mol
Siedepunkt: 155
Dichte: 1.05 g/cm³ (20 °C)
MDL: MFCD00000699
CAS-Nummer: 28788-62-7
EINECS: 249-224-2
UN: 3265

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR