2'-Hydroxy-4'-methylacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 1-(2-Hydroxy-4-methylphenyl)ethanone

BTB09999.250 BTB09999DA
MAYBBTB09999.250EA 0 EUR
MAYBBTB09999.250 MAYBBTB09999DA
2'-Hydroxy-4'-methylacetophenon
2'-Hydroxy-4'-methylacetophenon
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
MDL: MFCD00100559
CAS-Nummer: 6921-64-8
EINECS: 230-040-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR