4-(Ethylamino)benzoesäure
Lieferant: Thermo Scientific
BTB10315CB
BTB10315DA
MAYBBTB10315CBEA
0
EUR
MAYBBTB10315CB
MAYBBTB10315DA
4-(Ethylamino)benzoesäure
4-(Ethylamino)benzoic acid
Formel:
C₉H₁₁NO₂ |
MDL:
MFCD00102118 CAS-Nummer: 7409-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...