2,5-Dibromterephthalsäure

Lieferant: Thermo Scientific

Synonyme: 2,5-Dibromoterephtalic acid

BTB11564.250 BTB11564DA
MAYBBTB11564.250EA 0 EUR
MAYBBTB11564.250 MAYBBTB11564DA
2,5-Dibromterephthalsäure
2,5-Dibromterephthalsäure
Formel: C₈H₄Br₂O₄
Molecular Weight: 323.9200 g/mol
MDL: MFCD00204593
CAS-Nummer: 13731-82-3
EINECS: 237-300-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR