1-Hydroxy-9-fluorenon
Lieferant: CHEMPUR
BTB12320.250
BTB12320.1
CHMPBTB12320.250EA
0
EUR
CHMPBTB12320.250
MAYBBTB12320.1
1-Hydroxy-9-fluorenon
1-Hydroxy-9-fluorenon
Formel:
C₁₃H₈O₂ Molecular Weight: 196.21 g/mol Siedepunkt: 353 Schmelzpunkt: 115 Dichte: 1.39 g/cm³ (20 °C) |
CAS-Nummer:
6344-60-1 EINECS: 228-746-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...