2'-Hydroxy-4',6'-dimethoxyacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-on

BTB12407.1
MAYBBTB12407.1EA 0 EUR
MAYBBTB12407.1
2'-Hydroxy-4',6'-dimethoxyacetophenon
2'-Hydroxy-4',6'-dimethoxyacetophenon
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00017243
CAS-Nummer: 90-24-4
EINECS: 201-978-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR