(5-Mercapto-1,3,4-thiadiazol-2-ylthio)essigsäure
Lieferant: Thermo Scientific
Synonyme:
[(5-Sulphanyl-1,3,4-thiadiazol-2-yl)sulphanyl]acetic acid
BTB12604.10
MAYBBTB12604.10EA
0
EUR
MAYBBTB12604.10
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)essigsäure
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)essigsäure
Formel:
C₄H₄N₂O₂S₃ Molecular Weight: 208,28 g/mol |
MDL:
MFCD00129970 CAS-Nummer: 53723-88-9 EINECS: 258-728-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...