5-Brom-2-thenoesäure

Lieferant: Thermo Scientific

Synonyme: 5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure

BTB12643.1 BTB12643.10
MAYBBTB12643.1EA 0 EUR
MAYBBTB12643.1 MAYBBTB12643.10
5-Brom-2-thenoesäure
5-Brom-2-thenoesäure
Formel: C₅H₃BrO₂S
MDL: MFCD00079725
CAS-Nummer: 7311-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR