2-Brom-4'-pentylacetophenon

Lieferant: Thermo Scientific

Synonyme: 4-(Pent-1-yl)phenacyl bromide

BTB12841DA BTB12841EA
MAYBBTB12841DAEA 0 EUR
MAYBBTB12841DA MAYBBTB12841EA
2-Brom-4'-pentylacetophenon
2-Brom-4'-pentylacetophenon
Formel: C₁₃H₁₇BrO
CAS-Nummer: 64328-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR