4-Brom-2,5-dimethoxytoluol

Lieferant: Thermo Scientific

BTB13693.250
MAYBBTB13693.250EA 0 EUR
MAYBBTB13693.250
4-Brom-2,5-dimethoxytoluol
4-Brom-2,5-dimethoxytoluol
Formel: C₉H₁₁BrO₂
Molecular Weight: 231,09 g/mol
MDL: MFCD00666999
CAS-Nummer: 13321-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR