3-(Benzyloxy)-4,5-dimethoxybenzaldehyd
Lieferant: Thermo Scientific
BTB13918.250
BTB13918.1
MAYBBTB13918.250EA
0
EUR
MAYBBTB13918.250
MAYBBTB13918.1
3-(Benzyloxy)-4,5-dimethoxybenzaldehyd
3-(Benzyloxy)-4,5-dimethoxybenzaldehyd
Formel:
C₁₆H₁₆O₄ |
CAS-Nummer:
57230-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...