5-Chlor-o-kresol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Chlor-2-hydroxytoluol, 5-Chlor-2-methylphenol, 1-Chlor-3-hydroxy-4-methylbenzol
BTB14005.250
BTB14005.1
MAYBBTB14005.250EA
0
EUR
MAYBBTB14005.250
MAYBBTB14005.1
5-Chlor-o-kresol
5-Chlor-o-kresol
Formel:
C₇H₇ClO Molecular Weight: 142.58 g/mol Siedepunkt: 225 Schmelzpunkt: 75…76 °C |
MDL:
MFCD00060672 CAS-Nummer: 5306-98-9 EINECS: 226-160-3 UN: 3437 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...