5-Brom-1-indanon
Lieferant: CHEMPUR
Synonyme:
5-Bromo-2,3-dihydro-1H-inden-1-one
BTB14082.250
BTB14082.1
CHMPBTB14082.250EA
0
EUR
CHMPBTB14082.250
MAYBBTB14082.1
5-Brom-1-indanon
5-Brom-1-indanon
Formel:
C₉H₇BrO Siedepunkt: 280 °C (1013 hPa) Schmelzpunkt: 126…127 °C |
MDL:
MFCD00082718 CAS-Nummer: 34598-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...