(±)-Indolin-2-carbonsäure

Lieferant: Thermo Scientific

Synonyme: Indoline-2-carboxylic acid

BTB14322DA BTB14322EA
MAYBBTB14322DAEA 0 EUR
MAYBBTB14322DA MAYBBTB14322EA
(±)-Indolin-2-carbonsäure
(±)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
MDL: MFCD00010635
CAS-Nummer: 78348-24-0
EINECS: 278-899-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR