Bicyclo[3.3.1]-9-nonanon

Lieferant: CHEMPUR

Synonyme: Bicyclo[3.3.1]nonan-9-one

BTBG00160.250 BTBG00160.1
CHMPBTBG00160.250EA 0 EUR
CHMPBTBG00160.250 MAYBBTBG00160.1
Bicyclo[3.3.1]-9-nonanon
Bicyclo[3.3.1]-9-nonanon
Formel: C₉H₁₄O
Molecular Weight: 138.21
Schmelzpunkt: 156…158 °C
MDL: MFCD00074752
CAS-Nummer: 17931-55-4
EINECS: 241-868-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR