3,5-Dimethyl-4-isoxazolsulfonylchlorid

Lieferant: Thermo Scientific

CC00603.0001
MAYBCC00603.0001EA 0 EUR
MAYBCC00603.0001
3,5-Dimethyl-4-isoxazolsulfonylchlorid
3,5-Dimethyl-4-isoxazolsulfonylchlorid
Formel: C₅H₆ClNO₃S
Molecular Weight: 195,62 g/mol
MDL: MFCD00052554
CAS-Nummer: 80466-79-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR