(3,5-Dimethyl-4-isoxazolyl)methanol

Lieferant: Thermo Scientific

Synonyme: 3,5-Dimethyl-4-(hydroxymethyl)isoxazole

CC00609.250
MAYBCC00609.250EA 0 EUR
MAYBCC00609.250
(3,5-Dimethyl-4-isoxazolyl)methanol
(3,5-Dimethyl-4-isoxazolyl)methanol
Formel: C₆H₉NO₂
Molecular Weight: 127,1400 g/mol
MDL: MFCD00464226
CAS-Nummer: 19788-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR