4-Brom-3,5-dimethylisoxazol
Lieferant: Thermo Scientific
CC00610DA
CC00610.10
MAYBCC00610DAEA
0
EUR
MAYBCC00610DA
MAYBCC00610.10
4-Brom-3,5-dimethylisoxazol
4-Bromo-3,5-dimethylisoxazole
Formel:
C₅H₆BrNO Molecular Weight: 176,01 g/mol |
MDL:
MFCD00068187 CAS-Nummer: 10558-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...