3,5-Dimethyl-4-isoxazolboronsäure

Lieferant: Thermo Scientific

Synonyme: (dimethyl-1,2-oxazol-4-yl)boronic acid

CC00612.250
MAYBCC00612.250EA 0 EUR
MAYBCC00612.250
3,5-Dimethyl-4-isoxazolboronsäure
3,5-Dimethylisoxazole-4-boronic acid
Formel: C₅H₈BNO₃
Molecular Weight: 140,93 g/mol
MDL: MFCD02677945
CAS-Nummer: 16114-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR