3,4-Methylendioxyphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1,3-Benzodioxol-5-yl isothiocyanate

CC01607.250
MAYBCC01607.250EA 0 EUR
MAYBCC01607.250
3,4-Methylendioxyphenylisothiocyanat
3,4-Methylendioxyphenylisothiocyanat
Formel: C₈H₅NO₂S
MDL: MFCD00066321
CAS-Nummer: 113504-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR