3-Furancarbonsäure

Lieferant: Thermo Scientific

Synonyme: Furan-3-carboxylic acid, 3-Furancarboxylic acid

CC02101.50
MAYBCC02101.50EA 0 EUR
MAYBCC02101.50
3-Furancarbonsäure
3-Furoic acid
Formel: C₅H₄O₃
Molecular Weight: 112,08 g/mol
MDL: MFCD00005350
CAS-Nummer: 488-93-7
EINECS: 207-689-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR