4-Pyrid-4-ylbenzoesäure

Lieferant: Thermo Scientific

Synonyme: 4-Pyridin-4-yl-benzoic acid

CC035401DA
MAYBCC035401DAEA 0 EUR
MAYBCC035401DA
4-Pyrid-4-ylbenzoesäure
4-Pyrid-4-ylbenzoesäure
Formel: C₁₂H₉NO₂
Molecular Weight: 199,2100 g/mol
MDL: MFCD04114574
CAS-Nummer: 4385-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR