3-Pyrid-4-ylbenzoesäure

Lieferant: Thermo Scientific

Synonyme: 3-(Pyridin-4-yl)benzoic acid

CC035601DA
MAYBCC035601DAEA 0 EUR
MAYBCC035601DA
3-Pyrid-4-ylbenzoesäure
3-Pyrid-4-ylbenzoesäure
Formel: C₁₂H₉NO₂
Molecular Weight: 199,2054 g/mol
MDL: MFCD05864808
CAS-Nummer: 4385-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR