1-Methylimidazol-4-carbonsäure

Lieferant: Thermo Scientific

Synonyme: 1-Methyl-1H-imidazole-4-carboxylic acid

CC03601CB CC03601-0001
MAYBCC03601CBEA 0 EUR
MAYBCC03601CB MAYBCC03601-0001
1-Methylimidazol-4-carbonsäure
1-Methylimidazol-4-carbonsäure
Formel: C₅H₆N₂O₂
Molecular Weight: 126,11 g/mol
MDL: MFCD02179560
CAS-Nummer: 41716-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR