1-Methyl-4-imidazolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 1-Methyl-1H-imidazole-4-sulphonyl chloride

CC03603CB CC03603DA
MAYBCC03603CBEA 0 EUR
MAYBCC03603CB MAYBCC03603DA
1-Methyl-4-imidazolsulfonylchlorid
1-Methyl-4-imidazolsulfonylchlorid
Formel: C₄H₅ClN₂O₂S
Molecular Weight: 180,61 g/mol
MDL: MFCD00068060
CAS-Nummer: 137049-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR