Nicotinaldehyd

Lieferant: Thermo Scientific

Synonyme: Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde

CC04104.250
MAYBCC04104.250EA 0 EUR
MAYBCC04104.250
Nicotinaldehyd
Nicotinaldehyd
Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
MDL: MFCD00006382
CAS-Nummer: 500-22-1
EINECS: 207-900-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR