Isonicotinaldehyd

Lieferant: Thermo Scientific

CC04204.250 CC04204.1
MAYBCC04204.250EA 0 EUR
MAYBCC04204.250 MAYBCC04204.1
Isonicotinaldehyd
Isonicotinaldehyd
Formel: C₆H₅NO
MDL: MFCD00006425
CAS-Nummer: 872-85-5
EINECS: 212-832-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR