Isonicotinaldehyd
Lieferant: Thermo Scientific
CC04204.250
CC04204.1
MAYBCC04204.250EA
0
EUR
MAYBCC04204.250
MAYBCC04204.1
Isonicotinaldehyd
Isonicotinaldehyd
Formel:
C₆H₅NO |
MDL:
MFCD00006425 CAS-Nummer: 872-85-5 EINECS: 212-832-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...