2-Methyl-4-thiazolcarbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-Methylthiazole-4-carboxylic acid

CC05701CB CC05701DA CC05701-0001
MAYBCC05701CBEA 0 EUR
MAYBCC05701CB MAYBCC05701DA MAYBCC05701-0001
2-Methyl-4-thiazolcarbonsäure
2-Methyl-4-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,1637 g/mol
MDL: MFCD03407332
CAS-Nummer: 35272-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR