2-Benzofurancarbonsäure

Lieferant: CHEMPUR

Synonyme: Benzo[b]furan-2-carboxylic acid

CC06601.250 CC06601.1
CHMPCC06601.250EA 0 EUR
CHMPCC06601.250 MAYBCC06601.1
2-Benzofurancarbonsäure
Benzofuran-2-carboxylic acid
Formel: C₉H₆O₃
Molecular Weight: 162.15 g/mol
MDL: MFCD00005848
CAS-Nummer: 496-41-3
EINECS: 207-818-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR