(1H-Imidazol-2-yl)methanol Hydrochlorid

Lieferant: Thermo Scientific

Synonyme: 2-(Hydroxymethyl)-1H-imidazole hydrochloride

CC08909.250 CC08909DA
MAYBCC08909.250EA 0 EUR
MAYBCC08909.250 MAYBCC08909DA
(1H-Imidazol-2-yl)methanol Hydrochlorid
(1H-Imidazol-2-yl)methanol Hydrochlorid
Formel: C₄H₇ClN₂O
MDL: MFCD03659700
CAS-Nummer: 116177-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR