5-(Brommethyl)-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

CC09208CB CC09208DA
MAYBCC09208CBEA 0 EUR
MAYBCC09208CB MAYBCC09208DA
5-(Brommethyl)-2,1,3-benzothiadiazol
5-(Bromomethyl)-2,1,3-benzothiadiazole
Formel: C₇H₅BrN₂S
MDL: MFCD00544836
CAS-Nummer: 65858-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR