5-(Brommethyl)benzofurazan

Lieferant: Thermo Scientific

Synonyme: 5-(Bromomethyl)benzo[c][1,2,5]oxadiazole, 5-(Bromomethyl)-2,1,3-benzoxadiazole

CC09708CB CC09708DA
MAYBCC09708CBEA 0 EUR
MAYBCC09708CB MAYBCC09708DA
5-(Brommethyl)benzofurazan
5-(Brommethyl)benzofurazan
Formel: C₇H₅BrN₂O
Molecular Weight: 213,0314 g/mol
MDL: MFCD00174369
CAS-Nummer: 32863-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR