5-Isoxazolcarbonsäure

Lieferant: Thermo Scientific

Synonyme: Isoxazole-5-carboxylic acid

CC10301.250 CC10301DA
MAYBCC10301.250EA 0 EUR
MAYBCC10301.250 MAYBCC10301DA
5-Isoxazolcarbonsäure
5-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD00156151
CAS-Nummer: 21169-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR