3-(Hydroxymethyl)-5-methylisoxazol

Lieferant: Thermo Scientific

Synonyme: 3-Hydroxymethyl-5-methylisoxazole, 5-Methyl-3-isoxazolemethanol

CC10409DA
MAYBCC10409DAEA 0 EUR
MAYBCC10409DA
3-(Hydroxymethyl)-5-methylisoxazol
3-(Hydroxymethyl)-5-methylisoxazole
Formel: C₅H₇NO₂
Molecular Weight: 113,11 g/mol
MDL: MFCD00085129
CAS-Nummer: 35166-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR