Methyl-5-methylisoxazol-3-carboxylat

Lieferant: Thermo Scientific

Synonyme: Methyl 5-methylisoxazole-3-carboxylate

CC10422DA CC10422.50
MAYBCC10422DAEA 0 EUR
MAYBCC10422DA MAYBCC10422.50
Methyl-5-methylisoxazol-3-carboxylat
Methyl-5-methylisoxazol-3-carboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
MDL: MFCD00015895
CAS-Nummer: 19788-35-3
EINECS: 243-311-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR