2-(2-Thienyl)-4-thiazolcarbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-(2-Thienyl)-1,3-thiazole-4-carboxylicacid

CC11501-0001
MAYBCC11501-0001EA 0 EUR
MAYBCC11501-0001
2-(2-Thienyl)-4-thiazolcarbonsäure
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formel: C₈H₅NO₂S₂
Molecular Weight: 211,25 g/mol
MDL: MFCD01936014
CAS-Nummer: 24044-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR