3-Benzo[b]thiophencarbonylchlorid

Lieferant: Thermo Scientific

CC12302.250 CC12302DA
MAYBCC12302.250EA 0 EUR
MAYBCC12302.250 MAYBCC12302DA
3-Benzo[b]thiophencarbonylchlorid
3-Benzo[b]thiophencarbonylchlorid
Formel: C₉H₅ClOS
CAS-Nummer: 39827-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR