7-Brom-3,4-dihydro-2H-1,5-benzodioxepin

Lieferant: Thermo Scientific

CC13210DA CC13210EA
MAYBCC13210DAEA 0 EUR
MAYBCC13210DA MAYBCC13210EA
7-Brom-3,4-dihydro-2H-1,5-benzodioxepin
7-Brom-3,4-dihydro-2H-1,5-benzodioxepin
Formel: C₉H₉BrO₂
MDL: MFCD00508291
CAS-Nummer: 147644-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR