2-(3-Pyridinyl)-4-thiazolcarbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-(Pyridin-3-yl)thiazole-4-carboxylic acid

CC13701CB CC13701DA CC13701-0001
MAYBCC13701CBEA 0 EUR
MAYBCC13701CB MAYBCC13701DA MAYBCC13701-0001
2-(3-Pyridinyl)-4-thiazolcarbonsäure
2-(3-Pyridinyl)-4-thiazolcarbonsäure
Formel: C₉H₆N₂O₂S
Molecular Weight: 206,22 g/mol
MDL: MFCD00052304
CAS-Nummer: 39067-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR