2-Phenyl-4-thiazolcarbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-Phenylthiazole-4-carboxylic acid

CC18301CB CC18301DA
MAYBCC18301CBEA 0 EUR
MAYBCC18301CB MAYBCC18301DA
2-Phenyl-4-thiazolcarbonsäure
2-Phenyl-4-thiazolecarboxylic acid
Formel: C₁₀H₇NO₂S
Molecular Weight: 205,23 g/mol
MDL: MFCD00141954
CAS-Nummer: 7113-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR