4-(1H-Pyrazol-1-yl)benzoesäure

Lieferant: Thermo Scientific

CC18401.250
MAYBCC18401.250EA 0 EUR
MAYBCC18401.250
4-(1H-Pyrazol-1-yl)benzoesäure
4-(1H-Pyrazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,18 g/mol
MDL: MFCD02682056
CAS-Nummer: 16209-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR