4-(1H-1,2,4-Triazol-1-yl)benzoesäure

Lieferant: Thermo Scientific

Synonyme: 4-[1,2,4]Triazol-1-yl-benzoic acid

CC18601CB CC18601DA
MAYBCC18601CBEA 0 EUR
MAYBCC18601CB MAYBCC18601DA
4-(1H-1,2,4-Triazol-1-yl)benzoesäure
4-(1H-1,2,4-Triazol-1-yl)benzoesäure
Formel: C₉H₇N₃O₂
Molecular Weight: 189,1708 g/mol
MDL: MFCD02682060
CAS-Nummer: 162848-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR