4-(1H-1,2,4-Triazol-1-yl)benzaldehyd

Lieferant: Thermo Scientific

Synonyme: 1-(4-Formylphenyl)-1,2,4-triazole, 4-(1,2,4-Triazol-1-yl)benzaldehyde

CC18604.250 CC18604DA
MAYBCC18604.250EA 0 EUR
MAYBCC18604.250 MAYBCC18604DA
4-(1H-1,2,4-Triazol-1-yl)benzaldehyd
4-(1H-1,2,4-Triazol-1-yl)benzaldehyd
Formel: C₉H₇N₃O
Molecular Weight: 173,1714 g/mol
MDL: MFCD02681969
CAS-Nummer: 27996-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR