1-Methylimidazol-5-carbaldehyd

Lieferant: Thermo Scientific

Synonyme: 1-Methylimidazole-5-carboxaldehyde, 1-Methyl-1H-imidazole-5-carbaldehyde

CC23604.250 CC23604DA
MAYBCC23604.250EA 0 EUR
MAYBCC23604.250 MAYBCC23604DA
1-Methylimidazol-5-carbaldehyd
1-Methylimidazol-5-carbaldehyd
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
MDL: MFCD00030439
CAS-Nummer: 39021-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR