3-Bromimidazo[1,2-a]pyridin

Lieferant: Thermo Scientific

CC25410.250 CC25410DA
MAYBCC25410.250EA 0 EUR
MAYBCC25410.250 MAYBCC25410DA
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL: MFCD00014556
CAS-Nummer: 4926-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR